Learn More
Med Vet International IV Burette Set, 60Dr, J0468B
Catalog No. 502831803
Encompass_Preferred
This IV is for small precise volumes that can be accurately measured out in calibrated burette versus 1, 000ml IV fluid bags. Very useful for very small patients for accurate dosing. Luer Slip. Burette Capacity 100cc, 68"L.
Catalog No. 50-283-1803 Supplier Med Vet International Supplier No. JOR468B
This IV is for small precise volumes that can be accurately measured out in calibrated burette versus 1, 000ml IV fluid bags. Very useful for very small patients for accurate dosing. Luer Slip. Burette Capacity 100cc, 68"L.