Learn More
Bound Tree Medical Continu-Flo Non-Vented Minidrip IV Administration Set, 106in L, With 3 Interlink Injection Sites
Catalog No. NC0273983
Encompass_Preferred
Minidrip Continu-Flo solution set with 3 Interlink injection sites 6in, 42in and 86in from male luer lock adapter with retractable collar. Approximately 60 drops/mL . Approximate overall length 105in (2.7m). Set has tubing that is compatible with the colleague series volumetric infusion pumps and Flo-Gard™ 6201 and 6301 series volumetric infusion pumps. Contains DEHP. Fluid path is sterile, nonpyrogenic
Catalog No. NC0273983 Supplier Bound Tree Medical Supplier No. 356546
Minidrip Continu-Flo solution set with 3 Interlink injection sites 6in, 42in and 86in from male luer lock adapter with retractable collar. Approximately 60 drops/mL . Approximate overall length 105in (2.7m). Set has tubing that is compatible with the colleague series volumetric infusion pumps and Flo-Gard™ 6201 and 6301 series volumetric infusion pumps. Contains DEHP. Fluid path is sterile, nonpyrogenic