Learn More
Inorganic Ventures ICP CALIBRATION STD 27 125mL
SDP
Catalog No. 507527029
Encompass_Preferred
Inorganic Ventures' ICP-MS Calibration Standard 27 is a 125mL certified reference material set in a nitric acid matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-752-7029 Supplier Inorganic Ventures Supplier No. IVSTOCK27125ML
Inorganic Ventures' ICP-MS Calibration Standard 27 is a 125mL certified reference material set in a nitric acid matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search