Learn More
Inorganic Ventures ICP MS CALIBRATION STD 30 125
SDP
Catalog No. 507527026
Encompass_Preferred
Inorganic Ventures' ICP-MS Calibration Standard 30 is a 125mLcertified reference materialset in a nitric acid matrix for stability. This beryllium standard isTraceable to NISTand manufactured in accordance with our stringentQuality Assuranceguidelines. One yearExpiration Date.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-752-7026 Supplier Inorganic Ventures Supplier No. IVSTOCK30125ML
Inorganic Ventures' ICP-MS Calibration Standard 30 is a 125mLcertified reference materialset in a nitric acid matrix for stability. This beryllium standard isTraceable to NISTand manufactured in accordance with our stringentQuality Assuranceguidelines. One yearExpiration Date.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search