Learn More
Inorganic Ventures ICP MULTI ELEMENT STD 125mL
SDP
Supplier:  Inorganic Ventures IVSTOCK31125ML
Encompass_Preferred
Inorganic Ventures' ICP-MS Calibration Standard 31 is a 125mL certified reference material set in a nitric acid matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-752-7025