Learn More
Inorganic Ventures 100 ppm 8 Element ICP Calibration/Quality Control Standard, IV-STOCK-58-125ML
SDP
Catalog No. NC3584281
Encompass_Preferred
Part #: IV-STOCK-58-125ML, Description: 100 ppm 8 Element ICP Calibration/Quality Control Standard, Matrix: 10% v/v Hydrochloric Acid, Instrument(s): Common Multi-Element Standards
Catalog No. NC3584281 Supplier Inorganic Ventures Supplier No. IVSTOCK58125ML
Part #: IV-STOCK-58-125ML, Description: 100 ppm 8 Element ICP Calibration/Quality Control Standard, Matrix: 10% v/v Hydrochloric Acid, Instrument(s): Common Multi-Element Standards