Learn More
Med Vet International Oasis IV Mini Bore T-Connector, Sterile 6-7-in
Catalog No. 502522164
Encompass_Preferred
Distal end has a female luer fitting. Proximal end has male luer lock with latex free injection port. Also slide clamp for control. Sterile.
Catalog No. 50-252-2164 Supplier Med Vet International Supplier No. TPORTMB
 Restricted Item
Add to Cart
Add to Cart
Distal end has a female luer fitting. Proximal end has male luer lock with latex free injection port. Also slide clamp for control. Sterile.