Learn More
Description
- Trainees can practice a wide range of procedures, including venipuncture, IV cannulation, IV injection, blood collection with vacuum systems, infusion therapy, and butterfly catheter placement.
- Palpable veins and a realistic rolling vein on the hand enhance tactile feedback and help develop accurate technique.
- The soft, durable skin allows repeated punctures, while flexible fingers enable natural hand positioning during training.
- A pressurized blood reservoir ensures visible blood flashback for realistic simulation.
- The Closed-loop fluid system supports continuous use during infusion training.
- Exchangeable insertion sites (antecubital area and hand or forearm) can be replaced in under 60 seconds, making the trainer ideal for frequent and intensive use. It is compatible with standard clinical equipment, easy to clean, and comes with a carry bag for convenient storage and transport.
- The IV Injection Arm Trainer P51 is a practical and affordable solution for training essential vascular access skills
- Palpable veins and rolling vein for realistic training
- Visible blood flashback via pressurized reservoir
- Replaceable insertion sites for extended product life
- Compatible with standard clinical and vacuum systems
- Easy to clean and maintain
Specifications
Specifications
| Product Type | Manikin |
| For Use With (Application) | IV Injection Training |
| Color | Dark Skin |
| Gender | Male, Female |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.


