Learn More
Medicus Health IV Transport/Delivery System, 12 Hook Double Top
SDP
Catalog No. NC3353850
Encompass
IV Transport/Delivery System, 12 Hook Double Top features two stainless steel 1”-7/8” telescoping uprights that can adjust overall height of stand from 68”-85”
∙ IV Transport System includes a 6 outlet UL Hospital grade power strip with a 15′ cord
∙ IV Pole Delivery System has a capacity of up to 6 + Pumps (accommodate up to 9 devices)
∙ Innovative rake top design for improved line ID can hold up to 12 lines
∙ Six 4” twin wheel casters provide easy mobility
6 outlet Hospital grade power strip with a 15’ cord and cord wrap
∙ 25” diameter low center of gravity fabricated steel base
∙ Built-in powder coated handle to assist in steering
∙ Stainless Steel HD platform designed to accommodate multi-pumps
∙ Lower pole is powder coated 1.75” steel tubing
∙ Base weights 36 lbs. with total weight of pole at 60 lbs.
This item is not returnable. View return policy
Catalog No. NC3353850 Supplier Medicus Health Supplier No. 7505M1
IV Transport/Delivery System, 12 Hook Double Top features two stainless steel 1”-7/8” telescoping uprights that can adjust overall height of stand from 68”-85”
∙ IV Transport System includes a 6 outlet UL Hospital grade power strip with a 15′ cord
∙ IV Pole Delivery System has a capacity of up to 6 + Pumps (accommodate up to 9 devices)
∙ Innovative rake top design for improved line ID can hold up to 12 lines
∙ Six 4” twin wheel casters provide easy mobility
6 outlet Hospital grade power strip with a 15’ cord and cord wrap
∙ 25” diameter low center of gravity fabricated steel base
∙ Built-in powder coated handle to assist in steering
∙ Stainless Steel HD platform designed to accommodate multi-pumps
∙ Lower pole is powder coated 1.75” steel tubing
∙ Base weights 36 lbs. with total weight of pole at 60 lbs.