Learn More
Health Care Logistics Inc IV Tubing Covers, Roll, Amber
SDP
Catalog No. NC1808147
Encompass
UV Blocking - Amber IV Tubing Cover provides additional protection for light-sensitive medication. | Thickness - 1.5-mil | Dimensions: 2½"W x 300'L
Catalog No. NC1808147 Supplier Health Care Logistics Inc Supplier No. 19645
UV Blocking - Amber IV Tubing Cover provides additional protection for light-sensitive medication. | Thickness - 1.5-mil | Dimensions: 2½"W x 300'L