Learn More
Medchemexpress LLC Ivabradine-d6 hydrochloride | 2070009-63-9 | >96.26% | 5 MG
SDP
Catalog No. 5000406456
Encompass_Preferred
Ivabradine-d6 hydrochloride is a deuterium-labeled form of Ivabradine hydrochloride. It acts as a novel If inhibitor, functioning as a pure heart rate lowering agent. This compound is suitable for use as a tracer and as an internal standard.
  • Functions as a pure heart rate lowering agent
  • Suitable for use as a tracer
  • Ideal as an internal standard for quantitative analysis methods
Catalog No. 50-004-06456 Supplier Medchemexpress LLC Supplier No. HYB0162AS5MG
Ivabradine-d6 hydrochloride is a deuterium-labeled form of Ivabradine hydrochloride. It acts as a novel If inhibitor, functioning as a pure heart rate lowering agent. This compound is suitable for use as a tracer and as an internal standard.
  • Functions as a pure heart rate lowering agent
  • Suitable for use as a tracer
  • Ideal as an internal standard for quantitative analysis methods