Learn More
Apexbio Technology LLC Ivabradine HCl 148849-67-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000088713
Encompass_Preferred
Ivabradine hydrochloride (CAS 148849-67-6) is a selective inhibitor of the If (funny) current in sinoatrial node cells By targeting the If channel Ivabradine reduces cardiac pacemaker activity leading to a dose-dependent decrease in heart rate without affecting myocardial contractility In vitro studies report an IC50 of 2 9 M for inhibition of the If current Owing to its specific action on heart rate modulation Ivabradine HCl is widely utilized in preclinical research investigating cardiac electrophysiology autonomic regulation and the development of heart rate lowering agents
Catalog No. 50-000-88713 Supplier Apexbio Technology LLC Supplier No. B13605.1
Ivabradine hydrochloride (CAS 148849-67-6) is a selective inhibitor of the If (funny) current in sinoatrial node cells By targeting the If channel Ivabradine reduces cardiac pacemaker activity leading to a dose-dependent decrease in heart rate without affecting myocardial contractility In vitro studies report an IC50 of 2 9 M for inhibition of the If current Owing to its specific action on heart rate modulation Ivabradine HCl is widely utilized in preclinical research investigating cardiac electrophysiology autonomic regulation and the development of heart rate lowering agents