Learn More
Medchemexpress LLC Ivabradine hydrochloride | 148849-67-6 | 99.7% | 1 ML
SDP
Catalog No. 5000406254
Encompass_Preferred
Ivabradine hydrochloride is a potent and orally active HCN (hyperpolarization-activated cyclic nucleotide-gated) channel blocker. It inhibits the cardiac pacemaker current (If), reducing heart rate dose-dependently without modifying blood pressure. This compound demonstrates anticonvulsant, anti-ischaemic, and anti-anginal activities.
  • Potent and orally active HCN channel blocker
  • Inhibits cardiac pacemaker current (If)
  • Reduces heart rate dose-dependently
  • Exhibits anticonvulsant activity
  • Shows anti-ischaemic and anti-anginal effects
  • Investigated in clinical trials for conditions such as heart failure and atrial fibrillation
Catalog No. 50-004-06254 Supplier Medchemexpress LLC Supplier No. HYB0162A10MM/1ML
Ivabradine hydrochloride is a potent and orally active HCN (hyperpolarization-activated cyclic nucleotide-gated) channel blocker. It inhibits the cardiac pacemaker current (If), reducing heart rate dose-dependently without modifying blood pressure. This compound demonstrates anticonvulsant, anti-ischaemic, and anti-anginal activities.
  • Potent and orally active HCN channel blocker
  • Inhibits cardiac pacemaker current (If)
  • Reduces heart rate dose-dependently
  • Exhibits anticonvulsant activity
  • Shows anti-ischaemic and anti-anginal effects
  • Investigated in clinical trials for conditions such as heart failure and atrial fibrillation