Learn More
Medchemexpress LLC Ivabradine (hydrochloride) | 148849-67-6 | 99.7% | 100 MG
SDP
Catalog No. 5000406936
Encompass_Preferred
Ivabradine (hydrochloride) is a potent and orally active blocker of HCN (hyperpolarization-activated cyclic nucleotide-gated) channels. It inhibits the cardiac pacemaker current (If) and is known to reduce heart rate in a dose-dependent manner without affecting blood pressure. This compound exhibits anticonvulsant, anti-ischaemic, and anti-anginal activities, making it suitable for various research applications.
  • Potent and orally active HCN channel blocker
  • Inhibits cardiac pacemaker current (If)
  • Reduces heart rate dose-dependently without affecting blood pressure
  • Exhibits anticonvulsant activity
  • Exhibits anti-ischaemic activity
  • Exhibits anti-anginal activity
  • Intended for research and analytical applications
  • Purity of 99.7%
Catalog No. 50-004-06936 Supplier Medchemexpress LLC Supplier No. HYB0162A100MG
Ivabradine (hydrochloride) is a potent and orally active blocker of HCN (hyperpolarization-activated cyclic nucleotide-gated) channels. It inhibits the cardiac pacemaker current (If) and is known to reduce heart rate in a dose-dependent manner without affecting blood pressure. This compound exhibits anticonvulsant, anti-ischaemic, and anti-anginal activities, making it suitable for various research applications.
  • Potent and orally active HCN channel blocker
  • Inhibits cardiac pacemaker current (If)
  • Reduces heart rate dose-dependently without affecting blood pressure
  • Exhibits anticonvulsant activity
  • Exhibits anti-ischaemic activity
  • Exhibits anti-anginal activity
  • Intended for research and analytical applications
  • Purity of 99.7%