Learn More
Medchemexpress LLC HY-13017 10mg , Ivacaftor CAS:873054-44-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1301710MG
Encompass_Preferred
HY-13017 10mg Ivacaftor CAS: 873054-44-5 Ivacaftor (VX-770) is a potent and orally bioavailable CFTR potentiator, targeting G551D-CFTR and F508del-CFTR with EC50s of 100 nM and 25 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9060