Learn More
Medchemexpress LLC Ivacaftor-d9 | 1413431-07-8 | 99.8% | 401.55 g/mol | C24H19D9N2O3 | 5 MG
SDP
Catalog No. 5000263162
Encompass_Preferred
Ivacaftor-d9 is a deuterated analogue of ivacaftor used as a research CFTR potentiator. Supplied as a high-purity solid in a 5 mg pack, it is intended for analytical and pharmacological studies including mass spectrometry and metabolic stability experiments.
  • Deuterated analogue of ivacaftor for research applications.
  • CFTR potentiator activity used in cystic fibrosis research.
  • High purity suitable for analytical assays and bioanalytical work.
  • Solid form, convenient for storage and handling.
  • Useful as an isotopic internal standard for mass spectrometry.
  • Packaged in a small 5 mg quantity for experimental use.
Catalog No. 50-002-63162 Supplier Medchemexpress LLC Supplier No. HY13017S5MG
Ivacaftor-d9 is a deuterated analogue of ivacaftor used as a research CFTR potentiator. Supplied as a high-purity solid in a 5 mg pack, it is intended for analytical and pharmacological studies including mass spectrometry and metabolic stability experiments.
  • Deuterated analogue of ivacaftor for research applications.
  • CFTR potentiator activity used in cystic fibrosis research.
  • High purity suitable for analytical assays and bioanalytical work.
  • Solid form, convenient for storage and handling.
  • Useful as an isotopic internal standard for mass spectrometry.
  • Packaged in a small 5 mg quantity for experimental use.