Learn More
Selleck Chemical LLC Ivacaftor (VX-770) 50mg 873054-44-5
SDP
Catalog No. 508744
Encompass_Preferred
Ivacaftor (VX-770) is a selective potentiator of CFTR targeting G551D-CFTR and F508del-CFTR with EC50 of 100 nM and 25 nM in fisher rat thyroid cells, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-874-4 Supplier Selleck Chemical LLC Supplier No. S114450MG
Ivacaftor (VX-770) is a selective potentiator of CFTR targeting G551D-CFTR and F508del-CFTR with EC50 of 100 nM and 25 nM in fisher rat thyroid cells, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.