Learn More
Medchemexpress LLC Ivaltinostat (formic) | 98.3% | 473.56 | 1 MG
SDP
Catalog No. 5000391728
Encompass_Preferred
Ivaltinostat formic is an orally active, potent pan-HDAC inhibitor that features a hydroxamic acid moiety designed to bind zinc at the catalytic pocket. It inhibits deacetylation of histone H3 and tubulin, inducing p53 accumulation and promoting p53-dependent transactivation. This compound enhances the expression of MDM2 and p21 proteins, increases sensitivity of Gemcitabine-resistant cells to therapies, induces apoptosis, and demonstrates anti-tumour effects.
  • Potent pan-HDAC inhibitor
  • Inhibits deacetylation of histone H3 and tubulin
  • Promotes p53-dependent transactivation
  • Enhances expression of MDM2 and p21 proteins
  • Increases sensitivity of drug-resistant cells
  • Induces apoptosis
  • Exhibits anti-tumour effects
  • Attenuates oxidative stress and inflammation
Catalog No. 50-003-91728 Supplier Medchemexpress LLC Supplier No. HY16138A1MG
Ivaltinostat formic is an orally active, potent pan-HDAC inhibitor that features a hydroxamic acid moiety designed to bind zinc at the catalytic pocket. It inhibits deacetylation of histone H3 and tubulin, inducing p53 accumulation and promoting p53-dependent transactivation. This compound enhances the expression of MDM2 and p21 proteins, increases sensitivity of Gemcitabine-resistant cells to therapies, induces apoptosis, and demonstrates anti-tumour effects.
  • Potent pan-HDAC inhibitor
  • Inhibits deacetylation of histone H3 and tubulin
  • Promotes p53-dependent transactivation
  • Enhances expression of MDM2 and p21 proteins
  • Increases sensitivity of drug-resistant cells
  • Induces apoptosis
  • Exhibits anti-tumour effects
  • Attenuates oxidative stress and inflammation