Learn More
Medchemexpress LLC Ivarmacitinib | 1445987-21-2 | MFCD32701909 | 99.6% | 414.48 g/mol | C18H22N8O2S | 5 MG
SDP
Catalog No. 5000263112
Encompass_Preferred
Ivarmacitinib is a selective Janus kinase 1 (JAK1) inhibitor supplied as a small-molecule powder for research use. It inhibits JAK1-mediated STAT3 phosphorylation, induces apoptosis in target cells, and exhibits anti-proliferative and anti-inflammatory activity. The compound is intended for in vitro and preclinical studies probing JAK/STAT signaling and related immuno-inflammatory pathways.
  • Selective inhibition of JAK1-mediated signaling.
  • Demonstrated inhibition of STAT3 phosphorylation.
  • Supports studies of apoptosis, inflammation, and cell proliferation.
  • Available as a stable powder suitable for low-temperature storage.
  • Molecular weight 414.48 g/mol; formula C18H22N8O2S for reference.
  • Provided in small milligram quantities for research applications.
Catalog No. 50-002-63112 Supplier Medchemexpress LLC Supplier No. HY1127245MG
Ivarmacitinib is a selective Janus kinase 1 (JAK1) inhibitor supplied as a small-molecule powder for research use. It inhibits JAK1-mediated STAT3 phosphorylation, induces apoptosis in target cells, and exhibits anti-proliferative and anti-inflammatory activity. The compound is intended for in vitro and preclinical studies probing JAK/STAT signaling and related immuno-inflammatory pathways.
  • Selective inhibition of JAK1-mediated signaling.
  • Demonstrated inhibition of STAT3 phosphorylation.
  • Supports studies of apoptosis, inflammation, and cell proliferation.
  • Available as a stable powder suitable for low-temperature storage.
  • Molecular weight 414.48 g/mol; formula C18H22N8O2S for reference.
  • Provided in small milligram quantities for research applications.