Learn More
Innovive Inc IVC Mouse Lid, with Large Reemay® and Uniblend Filters for Enhanced Ventilation
Catalog No. NC2739933
Encompass
Minimum order quantity - 100. Order in increments of 100.For applications that require enhanced ventilation.Designed for use with the Innorack® IVC Mouse.For use with Innocage® mouse bottoms.100% PET plastic. BPA-Free.Recyclable.
Catalog No. NC2739933 Supplier Innovive Inc Supplier No. MVX6
Minimum order quantity - 100. Order in increments of 100.For applications that require enhanced ventilation.Designed for use with the Innorack® IVC Mouse.For use with Innocage® mouse bottoms.100% PET plastic. BPA-Free.Recyclable.