Learn More
Innovive Inc Rat Dual Filter Lid Tall
Catalog No. NC2167273
Encompass
Tall IVC rat lid with dual Reemay and Uniblend filters and external bottle cavity for use with tall wire bar in the Innorack Tall IVC Rat.
This item is not returnable. View return policy
Catalog No. NC2167273 Supplier Innovive Inc Supplier No. RVX8
Tall IVC rat lid with dual Reemay and Uniblend filters and external bottle cavity for use with tall wire bar in the Innorack Tall IVC Rat.