Learn More
Med Vet International Iverhart Max Large, Soft Chew, 50.1-100Lbs, 6 Month
Catalog No. 502827929
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Iverhart Max Soft Chew is for use in dogs eight weeks of age and older to prevent canine heartworm disease by eliminating the tissue stage of heartworm larvae (Dirofilaria immitis) for a month (30 days) after infection and for the treatment and control of roundworms (Toxocara canis, Toxascaris leonina), hookworms (Ancylostoma caninum, Uncinaria stenocephala, Ancylostoma braziliense), and tapeworms (Dipylidium caninum, Taenia pisiformis). See package insert for complete product details
Catalog No. 50-282-7929 Supplier Med Vet International Supplier No. RXIVERMAXLSC
 Restricted Item
Add to Cart
Add to Cart
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Iverhart Max Soft Chew is for use in dogs eight weeks of age and older to prevent canine heartworm disease by eliminating the tissue stage of heartworm larvae (Dirofilaria immitis) for a month (30 days) after infection and for the treatment and control of roundworms (Toxocara canis, Toxascaris leonina), hookworms (Ancylostoma caninum, Uncinaria stenocephala, Ancylostoma braziliense), and tapeworms (Dipylidium caninum, Taenia pisiformis). See package insert for complete product details