Learn More
Medchemexpress LLC HY-15310 500mg Medchemexpress, Ivermectin CAS:70288-86-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15310500MG
Encompass_Preferred
Medchemexpress, HY-15310 500mg Ivermectin CAS:70288-86-7 Ivermectin (MK-933) is a broad-spectrum anti-parasite agent. Ivermectin (MK-933) is a specific inhibitor of Impα/β1-mediated nuclear import and has potent antiviral activity towards both HIV-1 and dengue virus. It is a positive allosteric effector of P2X4 and the α7 neuronal nicotinic acetylcholine receptor (nAChRs). Ivermectin also inhibits bovine herpesvirus1 (BoHV-1) replication and inhibits BoHV-1 DNA polymerase nuclear import[2][3][4]. Ivermectin is a candidate therapeutic against SARS-CoV-2/COVID-19[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9116