Learn More
Selleck Chemical LLC Ivermectin 50mg 70288-86-7 MK933
SDP
Catalog No. 501361163
Encompass_Preferred
Ivermectin (MK-933, IVM) is a glutamate-gated chloride channel (GluCls) activator, used as a broad-spectrum antiparasitic drug. Ivermectin (MK-933, IVM) is a specific positive allosteric effector of P2X4 and ?7 nicotinic acetylcholine receptors (nAChRs). Ivermectin has potent antiviral activity towards both HIV-1 and dengue virus. Ivermectin induces autophagy through the AKT/mTOR signaling pathway and mitophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1163 Supplier Selleck Chemical LLC Supplier No. S135150MG
Ivermectin (MK-933, IVM) is a glutamate-gated chloride channel (GluCls) activator, used as a broad-spectrum antiparasitic drug. Ivermectin (MK-933, IVM) is a specific positive allosteric effector of P2X4 and ?7 nicotinic acetylcholine receptors (nAChRs). Ivermectin has potent antiviral activity towards both HIV-1 and dengue virus. Ivermectin induces autophagy through the AKT/mTOR signaling pathway and mitophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.