Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ivermectin, ≥95%, MP Biomedicals™

Catalog No. ICN19600991 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19600991 1 g
ICN19600983 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19600991 Supplier MP Biomedicals, Inc Supplier No. 0219600991
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ivermectin is a broad-spectrum antiparasitic agent. Ivermectin is a macrolide antiobiotic, & semisynthetic derivative of avermectin B1a and B1b

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70288-86-7
Molecular Formula C48H74O14
Molecular Weight (g/mol) 875.106
MDL Number MFCD00869511
InChI Key AZSNMRSAGSSBNP-RDKIRRGNSA-N
Synonym 22, 23-Dihydroavermectin B1, Hyvermectin
PubChem CID 122172888
SMILES CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C

Specifications

Type Ivermectin
Physical Form Crystalline Powder
Color White or White/Yellow
Quantity 1 g
Assay Percent Range ≥95%
Formula Weight 875.1
Chemical Name or Material Ivermectin
EINECSNumber 274-536-0
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.