Learn More
Cayman Chemical Ivermectin B1a, Purity: ≥95%, CAS: 71827-03-7, 25 mg
SDP
Catalog No. 502967808
Encompass_Preferred
The main component (>80%) of ivermectin; binds to glutamate-gated chloride channels, inducing irreversible channel opening; ivermectin inhibits replication of SARS-CoV-2 in Vero/hSLAM cells
Catalog No. 50-296-7808 Supplier Cayman Chemical Supplier No. 1876825 MG
The main component (>80%) of ivermectin; binds to glutamate-gated chloride channels, inducing irreversible channel opening; ivermectin inhibits replication of SARS-CoV-2 in Vero/hSLAM cells