Learn More
Apexbio Technology LLC Ivermectin B1a(Synonyms: Ivermectin, 22,23-Dihydroavermectin B1a, Avermectin B1a, MK-933, Stromectol, Mectizan), 5mg, CAS: 71827-03-7.
SDP
Catalog No. 5000061390
Encompass_Preferred
Ivermectin B1a (CAS 71827-03-7) is the primary constituent ( 80%) of the antiparasitic macrocyclic lactone ivermectin Its pharmacological effects originate from selective interaction with glutamate-gated chloride ion channels prevalent in neurons and pharyngeal muscles of nematodes This binding induces durable chloride influx causing sustained hyperpolarization or depolarization resulting in neuromuscular paralysis Ivermectin displays potent antiparasitic efficacy (EC 104 nM) and is clinically significant in treating human onchocerciasis reducing symptoms associated with the infection
Catalog No. 50-000-61390 Supplier Apexbio Technology LLC Supplier No. C39055
Ivermectin B1a (CAS 71827-03-7) is the primary constituent ( 80%) of the antiparasitic macrocyclic lactone ivermectin Its pharmacological effects originate from selective interaction with glutamate-gated chloride ion channels prevalent in neurons and pharyngeal muscles of nematodes This binding induces durable chloride influx causing sustained hyperpolarization or depolarization resulting in neuromuscular paralysis Ivermectin displays potent antiparasitic efficacy (EC 104 nM) and is clinically significant in treating human onchocerciasis reducing symptoms associated with the infection