Learn More
Medchemexpress LLC Ivermectin B1a | 71827-03-7 | >99.97% | 875.09 | 5 MG
SDP
Catalog No. 5000364575
Encompass_Preferred
Ivermectin B1a is a derivative of Avermectin B1a and a main component of Ivermectin. Ivermectin is a broad-spectrum anti-parasite agent and a candidate therapeutic against SARS-CoV-2/COVID-19. Ivermectin B1a is the major component of Ivermectin.
  • Derivative of Avermectin B1a
  • Main component of Ivermectin
  • Broad-spectrum anti-parasite agent
  • Candidate therapeutic against SARS-CoV-2/COVID-19
Catalog No. 50-003-64575 Supplier Medchemexpress LLC Supplier No. 5MGHY126937
Ivermectin B1a is a derivative of Avermectin B1a and a main component of Ivermectin. Ivermectin is a broad-spectrum anti-parasite agent and a candidate therapeutic against SARS-CoV-2/COVID-19. Ivermectin B1a is the major component of Ivermectin.
  • Derivative of Avermectin B1a
  • Main component of Ivermectin
  • Broad-spectrum anti-parasite agent
  • Candidate therapeutic against SARS-CoV-2/COVID-19