Learn More
Cayman Chemical IvermectIn B1a aglycon 1mg
SDP
Catalog No. 502982709
Encompass_Preferred
An acid degradation product produced by hydrolysis of the disaccharide unit of ivermectin; inhibits nematode larval development
Catalog No. 50-298-2709 Supplier Cayman Chemical Supplier No. 194421 MG
An acid degradation product produced by hydrolysis of the disaccharide unit of ivermectin; inhibits nematode larval development