Learn More
Cayman Chemical Ivermectin B1a aglycone, Purity: ≥99%, CAS: 73162-95-5, 5 mg
SDP
Catalog No. 502983054
Encompass_Preferred
An acid degradation product produced by hydrolysis of the disaccharide unit of ivermectin; inhibits nematode larval development
Catalog No. 50-298-3054 Supplier Cayman Chemical Supplier No. 194425 MG
An acid degradation product produced by hydrolysis of the disaccharide unit of ivermectin; inhibits nematode larval development