Learn More
Cayman Chemical IvermectIn B1b 2.5mg
SDP
Catalog No. 502994448
Encompass_Preferred
The minor component (<20%) of ivermectin; binds to glutamate-gated chloride channels, inducing irreversible channel opening; ivermectin inhibits replication of SARS-CoV-2 in Vero/hSLAM cells
Catalog No. 50-299-4448 Supplier Cayman Chemical Supplier No. 238242.5 MG
The minor component (<20%) of ivermectin; binds to glutamate-gated chloride channels, inducing irreversible channel opening; ivermectin inhibits replication of SARS-CoV-2 in Vero/hSLAM cells