Learn More
Medchemexpress LLC Ivermectin B1b | 70209-81-3 | MFCD00866559 | 98.7% | 861.07 | C47H72O14 | 500UG
SDP
Catalog No. 5000239125
Encompass_Preferred
Ivermectin B1b is the minor component of ivermectin, a semi-synthetic macrocyclic lactone used as an antiparasitic agent. Supplied for laboratory research, the compound is reported on the supplier page to inhibit replication of SARS-CoV-2 in cell culture. Typical analytical data listed for research packs include molecular formula C47H72O14, molecular weight 861.07 Da, and lot-dependent purity around 98.7%.
  • Minor component of ivermectin mixtures
  • Macrocyclic lactone with antiparasitic activity
  • Reported to inhibit SARS-CoV-2 replication in cell culture
  • Provided in small research pack sizes suitable for biochemical assays
  • Molecular formula C47H72O14 and molecular weight 861.07 Da
  • Typical purity about 98.7% (lot dependent)
Catalog No. 50-002-39125 Supplier Medchemexpress LLC Supplier No. HY125729500UG
Ivermectin B1b is the minor component of ivermectin, a semi-synthetic macrocyclic lactone used as an antiparasitic agent. Supplied for laboratory research, the compound is reported on the supplier page to inhibit replication of SARS-CoV-2 in cell culture. Typical analytical data listed for research packs include molecular formula C47H72O14, molecular weight 861.07 Da, and lot-dependent purity around 98.7%.
  • Minor component of ivermectin mixtures
  • Macrocyclic lactone with antiparasitic activity
  • Reported to inhibit SARS-CoV-2 replication in cell culture
  • Provided in small research pack sizes suitable for biochemical assays
  • Molecular formula C47H72O14 and molecular weight 861.07 Da
  • Typical purity about 98.7% (lot dependent)