Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leica Microsystems Ivesta 3 Stereo Microscope

Catalog No. 01672691 Shop All Leica Microsystems Products
Change view
Click to view available options
Head Style:
Binocular, 35° Inclined
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style
01-672-691 Binocular, 35° Inclined
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-691 Supplier Leica Microsystems Supplier No. 8122664
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Leica Ivesta 3 stereo microscope offers precise optics and user-friendly features, enhancing cell sorting tasks in labs. With advanced capabilities, it streamlines processes, providing reliable and efficient results. Elevate your workflow with the convenience of the Leica Ivesta 3 microscope.

The Leica Ivesta 3 stereo microscope is an ideal solution for routine cell sorting applications. Engineered with precision optics and user-friendly features, this microscope offers exceptional clarity and ease of use. With its advanced capabilities, the Leica Ivesta 3 streamlines routine cell sorting processes, providing researchers and professionals with a reliable and efficient tool for daily laboratory tasks. Elevate your workflow with the performance and convenience offered by the Leica Ivesta 3 stereo microscope.

  • 9:1 zoom with magnification 6.1x-55x (10x eyepieces, no supplementary lens) - provides field of view up to 37.6mm for fast, accurate inspection
  • 12 mm Depth of Field with Fusion Optics. Greenough optical design - provides very flat (plan) field of view and large depth of field
  • Complete apochromatic corrected microscopy system - 35° angle eye tubes - for comfort and convenience
  • 122 mm standard working distance (without lens shield) - ensures excellent accessibility to the samples
  • Resolution up to 250 lp/mm (without supplementary lens, 10x eyepieces), maximum resolution up to 500 lp/mm (2x supplementary lens, 10x eyepieces)
  • Antimicrobial PC/ABS housing with AgTreat™
  • Clickstops at 10x 20x 30x 40x and 50x - inspection at specific magnification can be easily repeated
  • Large range of supplementary lenses as well as eyepieces
Includes:
  • 10450983 Ivesta 3
  • 10447137 Eyepiece 10x/23B, adj., eyeglasses x2
  • 10446339 Focus column for S-Line
  • 10446340 Incident light base, small
  • 10446341 Sub base with reflector
  • 31120300 KL300 LED fiberoptic light source w/ps
  • 31120212 KL300 LED Bracket for routine focusdrive
  • 10446392 Universal light guide, 600 mm
  • 10447039 Dust cover antistatic, small
TRUSTED_SUSTAINABILITY

Specifications

Voltage 100/240 V
Magnification Power 6.1X to 55X
Objective Types Plan Apochromatic
Product Type Educational Microscope
Head Style Binocular, 35° Inclined
Lighting Type LED, Transmitted Illumination
Eyepiece WF 10X/23 mm, Adjustable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.