Learn More
Alkali Scientific IVF Culture Dish, 60 x 20mm, Treated, Sterile, 10/Sleeve, 500/cs
SDP
Supplier:  Alkali Scientific VF60
Encompass_Preferred
Whether working with sperm samples or oocytes, viability is paramount and sample handling is essential in IVF processes. Our high-quality dishes offer convenience and reliability for the collection and preparation of male and female gametes. These dishes offer perfect angle of reservoir well walls for locating and easy washing of samples as well as a true flat bottom for maintaining optimal temperature. They have an easy grip design for increased safety while handling. They are sterile, non-progenic, and produced under ISO 13485 with full lot traceability.
Catalog No. 50-238-8667