Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3713MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: IVRN/827. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: MCF-7 cells. Localized to upper spinous, granular layers in normal skin. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Purified involucrin from human keratinocytes. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 66-170kDa.Chromosome: 1q21.3. Gene Symbol: IVL. Gene ID: 3713. Ensemble: ENSG00000163207. Uniprot: NP_005538.
Catalog No. 50-188-4846