Learn More
Aviva Systems Biology Corp IVL Antibody - N-terminal region (ARP48675_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48675_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IVL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IVL. Alternate names: -ApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-014-53561