Learn More
Aviva Systems Biology Corp IVNS1ABP Antibody - N-terminal region (ARP33100_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33100_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against IVNS1ABP. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human IVNS1ABP. Alternate names: ND1, NS-1, NS1BP, FLARA3, KLHL39, NS1-BP, HSPC068ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-52889