Learn More
Medchemexpress LLC HY-18767 10mg Medchemexpress, Ivosidenib CAS:1448347-49-6 Purity:>98%
SDP
Catalog No. 502029270
Encompass_Preferred
Medchemexpress, HY-18767 10mg Ivosidenib CAS:1448347-49-6 Ivosidenib (AG-120) is an orally active inhibitor of isocitrate dehydrogenase 1 mutant (mIDH1) enzyme, it exhibits profound d-2-hydroxyglutatrate (2-HG) lowering in vivo. Ivosidenib (AG-120) has the potential for AML therapy due to its acceptable safety profile and clinical activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9270 Supplier Medchemexpress LLC Supplier No. HY1876710MG
Medchemexpress, HY-18767 10mg Ivosidenib CAS:1448347-49-6 Ivosidenib (AG-120) is an orally active inhibitor of isocitrate dehydrogenase 1 mutant (mIDH1) enzyme, it exhibits profound d-2-hydroxyglutatrate (2-HG) lowering in vivo. Ivosidenib (AG-120) has the potential for AML therapy due to its acceptable safety profile and clinical activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.