Learn More
Selleck Chemical LLC Ivosidenib
SDP
Catalog No. 5000000635
Encompass_Preferred
Ivosidenib (AG-120) is an orally available inhibitor of isocitrate dehydrogenase type 1 (IDH1) with potential antineoplastic activity
Catalog No. 50-000-00635 Supplier Selleck Chemical LLC Supplier No. S820625MG
Ivosidenib (AG-120) is an orally available inhibitor of isocitrate dehydrogenase type 1 (IDH1) with potential antineoplastic activity