Learn More
Cayman Chemical Ivermectin B1a monosaccharide, Purity: ≥99%, CAS: 71837-27-9, 5 mg
SDP
Catalog No. 502982590
Encompass_Preferred
Produced by selective hydrolysis of the terminal saccharide unit of ivermectin; inhibits nematode larval development
Catalog No. 50-298-2590 Supplier Cayman Chemical Supplier No. 194435 MG
Produced by selective hydrolysis of the terminal saccharide unit of ivermectin; inhibits nematode larval development