Learn More
Bio IVT IVTGRO CP MEDIUM 1 L
Catalog No. 502311084
Encompass
CP Medium; Celsis In Vitro Technologies; InVitroGRO; Antibiotic-free; Optimized; Longer shelf life; Sterile-filtered; Storage temp.: 4 deg.C; Used for plating hepatocytes; Size: 1L
Catalog No. 50-231-1084 Supplier Bio IVT Supplier No. Z990004
CP Medium; Celsis In Vitro Technologies; InVitroGRO; Antibiotic-free; Optimized; Longer shelf life; Sterile-filtered; Storage temp.: 4 deg.C; Used for plating hepatocytes; Size: 1L