Learn More
Selleck Chemical LLC IWP-2 10mg 686770-61-6
SDP
Supplier:  Selleck Chemical LLC S708510MG
Encompass_Preferred
IWP-2 is an inhibitor of Wnt processing and secretion with IC50 of 27 nM in a cell-free assay, selective blockage of Porcn-mediated Wnt palmitoylation, does not affect Wnt/?-catenin in general and displays no effect against Wnt-stimulated cellular responses. IWP-2 specifically inhibits CK1?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0736836