Learn More
BIOGEMS INTERNATIONAL INC IWP-2 1mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 68661671MG
Encompass
IWP-2 is a Wnt pathway inhibitor by inactivating the Porcupine O-acyltransferase which palmitoylates Wnt proteins. It is reported to inhibit embryonic stem cell self-renewal and promote cardiomyocyte differentiation from human stem cells.
Catalog No. 50-153-0240