Learn More
Abcam IWP-4, 10MG
Catalog No. 502682116
Encompass_Preferred
MW 496.6 Da, Purity >98%. Potent inhibitor of Wnt pathway regulatory protein porupine (Porcn). Blocks Wnt protein secretion and prevents the canonical and non-canonical signaling pathways. Blocks palmitylation, and subsquent secretion and activity of Wnt. Induces differentiation of cardiomyocytes from human ESCs and iPSCs.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2116 Supplier Abcam Supplier No. AB27807610MG
MW 496.6 Da, Purity >98%. Potent inhibitor of Wnt pathway regulatory protein porupine (Porcn). Blocks Wnt protein secretion and prevents the canonical and non-canonical signaling pathways. Blocks palmitylation, and subsquent secretion and activity of Wnt. Induces differentiation of cardiomyocytes from human ESCs and iPSCs.
The product is subject to the following: Abcam Restricted Use Statement