Learn More
Selleck Chemical LLC IWP-4-E0033-5MG
SDP
Supplier:  Selleck Chemical LLC E00335MG
Encompass_Preferred
IWP-4 is a potent Wnt/ -catenin signaling pathway inhibitor with an IC50 of 25 nM
Catalog No. 50-234-0808