Learn More
Medchemexpress LLC Iwp-4 | 686772-17-8 | MFCD04457754 | 99.8% | 496.62 g/mol | C23H20N4O3S3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY128795MG
Encompass_Preferred
IWP-4 is a small-molecule inhibitor of Wnt/β-catenin signaling that inactivates the acyltransferase Porcn, with an in vitro IC50 of 25 nM. Supplied as a solid for research use, it is used to block Wnt secretion and study Wnt-dependent processes in cell and developmental biology assays.
  • Potent Porcn inhibitor, IC50 = 25 nM.
  • Blocks Wnt secretion and downstream β-catenin signaling.
  • High chemical purity suitable for in vitro assays.
  • Available in multiple small-mass pack sizes for dosing flexibility.
  • Soluble in DMSO for cell-based and biochemical studies.
Catalog No. 50-208-1184