Learn More
Selleck Chemical LLC IWP-4-E0033-25MG
SDP
Catalog No. 503135615
Encompass_Preferred
IWP-4 is a potent Wnt/ -catenin signaling pathway inhibitor with an IC50 of 25 nM
Catalog No. 50-313-5615 Supplier Selleck Chemical LLC Supplier No. E003325MG
IWP-4 is a potent Wnt/ -catenin signaling pathway inhibitor with an IC50 of 25 nM