Learn More
STEMCELL Technologies IWP-4, Size: 1 mg
SDP
Catalog No. 501975941
Encompass
Inhibitor of WNT Production-4 (IWP-4) inhibits WNT signaling by inactivating Porcupine, a membrane-bound O-acyltransferase responsible for palmitoylating WNT proteins, which is essential for WNT signaling ability and secretion (Chen et al.). IWP-4 impairs WNT pathway activity in vitro with an IC₅₀ value of 25 nM (Chen et al.).DIFFERENTIATION· Promotes cardiomyocyte differentiation in human pluripotent stem cells after treatment with CHIR99021 (Lian et al., 2012, 2013; Sequiera et al.).· Promotes cardiomyocyte differentiation in human embryonic stem cells following primitive streak induction with BMP4 and Activin A (Hudson et al.).
Catalog No. 50-197-5941 Supplier STEMCELL Technologies Supplier No. 72552
Add to Cart
Add to Cart
Inhibitor of WNT Production-4 (IWP-4) inhibits WNT signaling by inactivating Porcupine, a membrane-bound O-acyltransferase responsible for palmitoylating WNT proteins, which is essential for WNT signaling ability and secretion (Chen et al.). IWP-4 impairs WNT pathway activity in vitro with an IC₅₀ value of 25 nM (Chen et al.).DIFFERENTIATION· Promotes cardiomyocyte differentiation in human pluripotent stem cells after treatment with CHIR99021 (Lian et al., 2012, 2013; Sequiera et al.).· Promotes cardiomyocyte differentiation in human embryonic stem cells following primitive streak induction with BMP4 and Activin A (Hudson et al.).