Learn More
Medchemexpress LLC IWP-O1 | 2074607-48-8 | 99.8% | 432.48 g/mol | C26H20N6O | 1 ML
SDP
Catalog No. 5000212319
Encompass_Preferred
IWP-O1 is a potent porcupine (Porcn) inhibitor used for research to block Wnt protein secretion and downstream signaling, with cellular activity reported in the picomolar range. It is supplied as a 10 mM solution in DMSO or as solid formats and includes documentation for handling and quality verification.
  • Potent porcupine inhibitor that blocks wnt protein secretion and signaling.
  • Demonstrated EC50 of 80 pM in L-Wnt-STF cellular assay.
  • Available as 10 mM solution in DMSO or as solid formats for flexibility.
  • High purity suitable for research use and analytical applications.
  • Characterized by formula C26H20N6O and molecular weight 432.48 g/mol.
  • Includes SDS and COA for safe handling and quality verification.
Catalog No. 50-002-12319 Supplier Medchemexpress LLC Supplier No. HY10085310MM 1ML
IWP-O1 is a potent porcupine (Porcn) inhibitor used for research to block Wnt protein secretion and downstream signaling, with cellular activity reported in the picomolar range. It is supplied as a 10 mM solution in DMSO or as solid formats and includes documentation for handling and quality verification.
  • Potent porcupine inhibitor that blocks wnt protein secretion and signaling.
  • Demonstrated EC50 of 80 pM in L-Wnt-STF cellular assay.
  • Available as 10 mM solution in DMSO or as solid formats for flexibility.
  • High purity suitable for research use and analytical applications.
  • Characterized by formula C26H20N6O and molecular weight 432.48 g/mol.
  • Includes SDS and COA for safe handling and quality verification.