Learn More
Selleck Chemical LLC IWP-O1
SDP
Catalog No. 5000001653
Encompass_Preferred
IWP-O1 is a porcn inhibitor with an EC50 value of 80 pM in a cultured cell reporter assay of Wnt signaling and effectively suppresses the phosphorylation of Dvl2/3 in HeLa cells
Catalog No. 50-000-01653 Supplier Selleck Chemical LLC Supplier No. S86455MG
IWP-O1 is a porcn inhibitor with an EC50 value of 80 pM in a cultured cell reporter assay of Wnt signaling and effectively suppresses the phosphorylation of Dvl2/3 in HeLa cells